Onguard™ Polyblend™ Work Shoes and Work Boots

Ideal for use in chemical and poultry processing plants, in mining, and in commercial fishing

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 861011100

 View more versions of this product

Catalog No. 17-986-321

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-05
    Closure Type Pull-on
    Color Black
    For Use With (Application) Used in lawn care, poultry and food processing, mining, breweries, oil rigs, oil refineries, and chemical plants
    Gender Male
    Height (English) 16 in.
    Lug Type Kick off
    Material Polyurethane, PVC
    Resistant Fat, Acid, Oil, Grease, chemicals
    Shank Material Steel
    Size 11
    Sole Features Gray cleated outsole, slip resistant , self cleaning soles
    Toe Material Plain
    Toe Style Plain

    Polyurethane and PVC plastic provide the moderate chemical resistance of these injection-molded, one-piece boots and work shoes.

    • Steel shanks
    • Removable, washable cushion insoles
    • Pull-up and kick-off lugs
    • Molded pant stays to keep pants tucked inside the boot shaft
    • Work shoes lace up
    • All styles are black with gray cleated nonslip outsoles

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit