Onguard™ Polyblend™ Work Shoes and Work Boots

Ideal for use in chemical and poultry processing plants, in mining, and in commercial fishing

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 861011200

 View more versions of this product

Catalog No. 17-986-322

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Closure Type Pull-on
    Height (English) 16 in.
    Sole Features Gray cleated outsole, slip resistant , self cleaning soles
    Toe Material Plain
    Color Black
    Size 12
    Certifications/Compliance ASTM F2413-05
    Gender Male
    Lug Type Kick off
    Resistant Fat, Acid, Oil, Grease, chemicals
    Shank Material Steel
    Toe Style Plain
    Material Polyurethane, PVC
    For Use With (Application) Used in lawn care, poultry and food processing, mining, breweries, oil rigs, oil refineries, and chemical plants

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit