Learn More
Onguard™ Polyblend™ Work Shoes and Work Boots
Ideal for use in chemical and poultry processing plants, in mining, and in commercial fishing
Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 861020800
 View more versions of this product
Catalog No. 17-986-328
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM F2413-05
Used in lawn care, poultry and food processing, mining, breweries, oil rigs, oil refineries, and chemical plants
16 in.
Fat, Acid, Oil, Grease, chemicals
Gray cleated outsole, slip resistant , self cleaning soles
Black
8
Pull-on
Male
Kick off
Steel
Steel
Polyurethane, PVC