Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Polymax™ Chemical Resistant Work Boots with Plain Toe
SDP

Ideal for hazardous waste management and chemical or food processing plants

Specifications

Certifications/Compliance ASTM F2413-05
Color Orange, Beige
Material PVC
For Use With (Application) Ideal for hazardous waste management and chemical or food processing plants
Height (English) 16 in.
View More Specs

Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-806-596
SDP
View Documents
Dunlop™
84075
15 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-084073
SDP
View Documents
Dunlop™
840751400
14 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-084070
SDP
View Documents
Dunlop™
840751100
11 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-084067
SDP
View Documents
Dunlop™
840750800
8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-084065
SDP
View Documents
Dunlop™
840750600
6 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-084072
SDP
View Documents
Dunlop™
840751300
13 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-084074
SDP
View Documents
Dunlop™
840751500
15 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • High-molecular-weight resins in these boots provide the same protection as more expensive neoprene
  • Ultragrip™ outsoles provide excellent traction and slip resistance
  • 16 in. high (41cm) with cut-off lines on the uppers at 12 and 14 in. (30.5 and 36cm)
  • Feature steel toes and shanks, removable molded preformed three-layer insoles, toe and heel bumpers, kick-off lugs, and reinforced around the ankle area for additional protection
  • Brown with beige outsoles

Compliance

Steel toe meets or exceeds ASTM™ F2413-11 specifications

Specifications

Specifications

ASTM F2413-05
PVC
16 in.
Seamless
Ultragrip™ sipe outsole, removable molded preformed three layer insoles
Orange, Beige
Ideal for hazardous waste management and chemical or food processing plants
Chemicals
Steel
Plain
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit