Learn More
Onguard™ Polymax™ Chemical Resistant Work Boots with Plain Toe
Ideal for hazardous waste management and chemical or food processing plants
Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 840751400
 View more versions of this product
Catalog No. 19-084073
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM F2413-05
Male
Chemical
Steel
Plain
PVC
Ideal for hazardous waste management and chemical or food processing plants
16 in.
Seamless
Ultragrip™ sipe outsole, removable molded preformed three layer insoles
Rust with Beige
14