Onguard™ Polymax™ Chemical Resistant Work Boots with Steel Toes

Ideal for hazardous waste management and chemical or food processing plants

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 840761400

 View more versions of this product

Catalog No. 17-986-312

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-05
    For Use With (Application) Ideal for hazardous waste management and chemical or food processing plants
    Gender Male
    Height (English) 16 in.
    Resistant Chemical
    Seam Type Seamless
    Shank Material Steel
    Sole Features Ultragrip™ sipe outsole, removable molded preformed three layer insoles
    Toe Material Steel
    Color Rust with Beige
    Material PVC
    Size 14

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit