Onguard™ Polymax™ Chemical Resistant Work Boots with Steel Toes

Ideal for hazardous waste management and chemical or food processing plants

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 840760900

 View more versions of this product

Catalog No. 17-986-307

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Ideal for hazardous waste management and chemical or food processing plants
    Gender Male
    Height (English) 16 in.
    Resistant Chemical
    Seam Type Seamless
    Toe Material Steel
    Size 9
    Certifications/Compliance ASTM F2413-05
    Shank Material Steel
    Sole Features Ultragrip™ sipe outsole, removable molded preformed three layer insoles
    Material PVC
    Color Rust with Beige

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit