Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dunlop™ Onguard™ Polymax™ Chemical Resistant Work Boots with Steel Toes
Ideal for hazardous waste management and chemical or food processing plants
Supplier: Dunlop™ 840761100
This item is not returnable.
View return policy
Description
- High-molecular-weight resins in these boots provide the same protection as more expensive neoprene
- Ultragrip™ outsoles provide excellent traction and slip resistance
- 16 in. high (41cm) with cut-off lines on the uppers at 12 and 14 in. (30.5 and 36cm)
- Feature steel toes and shanks, removable molded preformed three-layer insoles, toe and heel bumpers, kick-off lugs, and reinforced around the ankle area for additional protection
- Brown with beige outsoles
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications
Specifications
ASTM F2413-05 | |
Ideal for hazardous waste management and chemical or food processing plants | |
PVC | |
Steel | |
Ultragrip™ sipe outsole, removable molded preformed three layer insoles | |
Chemicals |
Orange, Beige | |
16 in. | |
Seamless | |
11 | |
Steel |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction