Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dunlop™ Onguard™ Pullover Slicker Boots
Fit easily over work shoes
Supplier: Dunlop™ 880501400
This item is not returnable.
View return policy
Description
- Waterproof with excellent cold weather flexibility and chemical resistance
- Nonconductive adjustable polyester strap with plastic buckle and snaps
- Nonabsorbent polyester lining
- 17 in. high (43cm)
- Yellow with black cleated soles and kick-off lugs

Specifications
Plastic | |
Construction, concrete work, utilities and food processing | |
Polyester | |
14 | |
Polyester |
Yellow | |
17 in. | |
Flexothane, PVC | |
Black self-cleaning cleated outsole | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction