Learn More
Dunlop™ Onguard™ Pullover Slicker Boots
SDP
Fit easily over work shoes
Supplier:  Dunlop™ 880500900
 View more versions of this product
Catalog No. 19-024528
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Waterproof with excellent cold weather flexibility and chemical resistance
  • Nonconductive adjustable polyester strap with plastic buckle and snaps
  • Nonabsorbent polyester lining
  • 17 in. high (43cm)
  • Yellow with black cleated soles and kick-off lugs
Specifications
Specifications
Plastic
Construction, concrete work, utilities and food processing
Polyester
9
Polyester
Yellow
17 in.
Flexothane, PVC
Black self-cleaning cleated outsole
Yes
Product Suggestions
Product Suggestions