Onguard™ Pullover Slicker Boots

Fit easily over work shoes

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 88050

 View more versions of this product

Catalog No. 19-821-977

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Construction, concrete work, utilities and food processing
    Height (English) 17 in.
    Sole Features Black self cleaning cleated outsole
    Strap Material Polyester
    Waterproof Yes
    Material PVC, Flex-O-Thane
    Buckle Material Plastic
    Gender Male
    Liner Material Polyester
    Color Yellow
    Size 17

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit