Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Onguard Sanitex Bib Overall

Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 71250MD00

 View more versions of this product

Catalog No. 19-120-893B


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ASTM D6413
Take up snap
127cm
PVC, Nylon, Polyester
Medium
Sanitex
Chemical plants, refineries, food processing and environmental clean-up
Adjustable snap
73.6cm
Heavy duty zipper
Green
50 in.
1.0
46 in.
Adjustable elastic with deluxe clip
0.35mm
29 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit