Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Onguard Sanitex Bib Overall

Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 712504X00

 View more versions of this product

Catalog No. 19-120-893G


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ASTM D6413
Take up snap
147.3cm
PVC, Nylon, Polyester
4X-Large
Sanitex
Chemical plants, refineries, food processing and environmental clean-up
Adjustable snap
83.8cm
Heavy duty zipper
Green
58 in.
1.0
66 in.
Adjustable elastic with deluxe clip
0.35mm
33 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit