Learn More
Dunlop™ Onguard™ Sanitex Coverall with Attached Hood
Manufacturer:  Dunlop™ 71220XL00
 View more versions of this product
Catalog No. 19-120-891D
Add to cart
Specifications
Specifications
ASTM D6413
Green
Attached
Elastic adjustable
Hooded
Nylon Polyester
PVC
X-Large
Chemical plants, refineries, food processing and environmental clean-up
Zipper
Attached hood, elastic adjustable suspender with deluxe suspender clip, cord locks on hood strings, adjustable waist snap, ankle and cuff take up snap for a snug/tapered fit