Onguard Sanitex Coverall with Attached Hood

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 712202X00

 View more versions of this product

Catalog No. 19-120-891E

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM D6413
    Color Green
    For Use With (Application) Chemical plants, refineries, food processing and environmental clean-up
    Material Nylon Polyester
    Includes Attached hood, elastic adjustable suspender with deluxe suspender clip, cord locks on hood strings, adjustable waist snap, ankle and cuff take up snap for a snug/tapered fit
    Size 2X-Large
    Coating PVC
    Closure Type Zipper
    Suspender Elastic adjustable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit