Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Standard Work Boots and Shoes
SDP

Ideal for wet, muddy areas where extreme chemical resistance is not necessary

Specifications

Certifications/Compliance ASTM
Color Black
For Use With (Application) For wet, muddy
Material PVC
Shank Material Steel
View More Specs

Products 8
Catalog Number Mfr. No. Size Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size Height (English) Price Quantity & Availability  
17-986-347
SDP
View Documents
Dunlop™
863121100
11 16 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
17-986-345
SDP
View Documents
Dunlop™
863120900
9 16 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
17-986-350
SDP
View Documents
Dunlop™
863040600
6 6 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
17-986-351
SDP
View Documents
Dunlop™
863040700
7 6 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
17-986-355
SDP
View Documents
Dunlop™
863041100
11 6 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
17-986-353
SDP
View Documents
Dunlop™
863040900
9 6 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
17-986-356
SDP
View Documents
Dunlop™
863041200
12 6 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
17-986-357
SDP
View Documents
Dunlop™
863041300
13 6 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

  • Excellent for construction and general maintenance
  • One-piece injection-molded boots provide waterproof protection, with steel toes, steel shanks, and cushion insoles.
  • Boots feature pull-up and kick-off lugs, cleated soles and cut-off lines (Cut boot shaft at cut-off line to shorten)
  • Lace-up work shoes have steel toes and steel shanks

Order Info

Also available: 16 in. high (41cm), plain toe; 16 in. high (41cm), white, steel toe; and 14 in. high (36cm), women's, black (cleated sole), plain or steel toe. Call your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information.

Compliance

Steel toes of boots and shoes meet or exceed ASTM™ F2413-11 specifications

Specifications

Specifications

ASTM
For wet, muddy
Steel
Standard
Yes
Black
PVC
Cleated outsole
Steel
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit