Onguard™ Standard Work Boots and Shoes

Ideal for wet, muddy areas where extreme chemical resistance is not necessary

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 863040800

 View more versions of this product

Catalog No. 17-986-352

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) For wet, muddy
    Gender Male
    Height (English) 6 in.
    Shank Material Steel
    Style Standard
    Toe Material Steel
    Color Black
    Material PVC
    Size 8
    Certifications/Compliance ASTM
    Sole Features Cleated outsole
    Waterproof Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit