Onguard™ Steel Midsole Work Boots

Ideal for demolition crews and recycling plants

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 868021000

 View more versions of this product

Catalog No. 19-024513

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM
    Gender Male
    Sole Features Ultragrip™ sole, full Steel midsole
    Style Standard
    Toe Material Steel
    Waterproof Yes
    Color Black
    Material Polyblend PVC
    Size 10
    For Use With (Application) Demolition crews and recycling plants
    Height (English) 16 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit