Onguard™ Steeltex™ Waterproof Apparel
Strong-as-steel protection for a wide range of applications
Manufacturer: Onguard Industries LLCSupplier Diversity Partner 75534 XL 00
 View more versions of this product
Catalog No. 17-977-438
  Check Availability
Add to Cart
    Specifications
    Coating Polyurethane
    For Use With (Application) For use in food processing, petrochemical processing, and general industy
    Size X-Large
    Includes Attached hood
    Material Polyurethane on heavy-duty nylon
    Hood Type Attached
    Liner Material Nylon
    Ideal for food processing, petrochemical processing, and industrial use.
    • Thin, lightweight, durable jacket with attached hood
    • Constructed of heavy-duty polyurethane on nylon for superior protection
    • Heat-welded seams for additional durability
    • Secure-fit snaps provide a sure fit
    • Offer excellent waterproof and moderate chemical protection
  • Description & Specifications
Related Products
Product Content Feedback