Learn More
Dunlop™ Onguard™ Superpoly Boots with Chevron Sole
Ideal for use in food processing, especially red-meat processing and petrochemical applications
Manufacturer:  Dunlop™ 853240400
 View more versions of this product
Catalog No. 19-064364
Add to cart
Description
Description
  • Waterproof with a high degree of chemical resistance
  • Steel toe and shank provide optimal protection
  • Chevron outsole provides excellent slip resistance
  • Pull-up and kick-off lugs
  • Removable or replaceable preformed insole for added comfort
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications
Specifications
Specifications
Chemicals
ASTM F2413-05, ANSI Z41 PT99 M I/75 C/75 PR
Pull-on
16 in.
PVC, Urethane
4
Steel
Safety
Red
Used in slaughter houses, red meat processing, canneries, breweries, dairies, oil rigs and chemical plants
Kick-off
Steel
Tan Chevron outsole and replaceable cushion insoles