Onguard™ SureFlex™ PVC Work Boots

Offer superior traction with high chemical and abrasion resistance

Manufacturer: Onguard Industries LLC 87982060

 View all options for this product

Catalog No. 19-087-147B

  Check Availability
Add to Cart


    Certifications/Compliance ASTM F2413-05
    Closure Type Pull-on
    Color Black
    For Use With (Application) Used in chemicals, farming and food processing
    Gender Male
    Height (English) 16 in.
    Liner Material Nylon
    Lug Type Kick off
    Material PVC
    Resistant Animal Fats, Petroleum, Oils, Grease, chemicals
    Shank Material Steel
    Size 6
    Sole Features Orange Safety-Loc™ Nitrile outsole, removable EVA cushion insole
    Toe Material Steel

    Soft, flexible PVC-compound upper gives boots a comfortable, rubber-like fit and feel. PVC/nitrile sole is 100% waterproof. Removable cushion insole absorbs perspiration to prevent odor and reduces foot discomfort.

    • PVC offers superior durability with excellent resistance to oils, chemicals, and fats
    • Orange Safety-Loc™ outsole grabs slippery surfaces for maximum traction
    • Reinforced steel shank provides extra arch support
    • Nylon lining and kick-off lugs allow easy on and off
    • Steel toe for optimal impact and crush resistance
    • 15 in. high (38.1cm)
    • Made in U.S.A.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit