Onguard™ SureFlex™ PVC Work Boots

Offer superior traction with high chemical and abrasion resistance

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 87982070

 View more versions of this product

Catalog No. 19-087-147C

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-05
    Gender Male
    Lug Type Kick off
    Shank Material Steel
    Closure Type Pull-on
    For Use With (Application) Used in chemicals, farming and food processing
    Height (English) 16 in.
    Liner Material Nylon
    Resistant Animal Fats, Petroleum, Oils, Grease, chemicals
    Sole Features Orange Safety-Loc™ Nitrile outsole, removable EVA cushion insole
    Toe Material Steel
    Color Black
    Material PVC
    Size 7

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit