Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Onguard Tuftex Bib Overall

Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 780502X00

 View more versions of this product

Catalog No. 19-120-918E


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ASTM D6413
Take up snap
142.2cm
PVC, Nylon, PVC
2X-Large
Tuftex
All-purpose use
Adjustable snap
81.2cm
Plain
Yellow
56 in.
2.0
58 in.
Adjustable
0.3mm
32 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit