Learn More
Dunlop™ Onguard™ Tuftex Three-Piece Suits
Lightweight, high-performance material with excellent tear resistance
Manufacturer:  Dunlop™ 780172X00
 View more versions of this product
Catalog No. 19-120-916E
Add to cart
Description
Description
Materials:
  • Yellow PVC, nylon and PVC scrim construction
  • 0.30mm thickness
  • Lightweight but very tear resistant
Jacket Features:
  • Storm flap front
  • Two front snap pockets
  • Underarm vents to keep you cool
Benefits:
  • Heavy duty adjustable suspender
  • Adjustable waist snaps for the perfect fit
  • Ankle and cuff take-up snaps for a snug/tapered fit
Specifications
Specifications
2.0
Adjustable
Yellow
2X-Large