Learn More
Dunlop™ Onguard™ Tuftex Three-Piece Suits
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
Description
Materials:
- Yellow PVC, nylon and PVC scrim construction
- 0.30mm thickness
- Lightweight but very tear resistant
- Storm flap front
- Two front snap pockets
- Underarm vents to keep you cool
- Heavy duty adjustable suspender
- Adjustable waist snaps for the perfect fit
- Ankle and cuff take-up snaps for a snug/tapered fit
Specifications
Color | Yellow |
Material | Nylon/PVC |
Size | 4X-Large |
No. of Pockets | 2.0 |