Onguard Visitex II Jackets with Detachable Hood
Manufacturer: Onguard Industries LLCSupplier Diversity Partner 721353X00
 View more versions of this product
Catalog No. 19-806-560
  Check Availability
Add to Cart
    Specifications
    Size 3X-Large
    Includes Detachable hooded cape-back Jacket with vents, ankle and cuff take up snap, cord locks on hood strings, suspender with deluxe suspender clip
    Material Polyester
    Tape Type 2 in. Reflective stripes
    Closure Type Storm Flap Front
    No. of Pockets 2.0
    Coating PVC
    Certifications/Compliance ASTM D6413
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback