Learn More
Dunlop™ Onguard™ Visitex II Jackets with Detachable Hood
Click to view available options
Size:
Medium
Large
2X-Large
3X-Large
Specifications
Size | Medium |
Material | Polyester |
Color | Yellow |
Closure Type | Storm Flap Front |
Certifications/Compliance | ASTM D6413 |
Coating | PVC |
No. of Pockets | 2 |
Tape Type | 2 in. Reflective stripes |