Onguard Waterproof Gator Overshoes

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 898100 LG

 View more versions of this product

Catalog No. 19-099-183C

  Check Availability
Add to Cart

    Specifications

    Closure Type Button, Elastic loops with stitched tabs
    For Use With (Application) Farming and utilities
    Height (English) 5 in.
    Sole Features Self-cleaning large lug outsole
    Waterproof Yes
    Color Black
    Material PVC
    Size Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit