Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Onguard Webtex Bib Overalls

Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 76052MD00

 View more versions of this product

Catalog No. 19-120-906B


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Heavy duty zipper, snap fly
Yellow
127cm
PVC, Polyester
Medium
Webtex
Designed for use in warm or cool climates
29 in.
Take up snap
PVC on non-woven Polyester
50 in.
2.0
46 in.
Adjustable
Adjustable snap
73.6cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit