Learn More
This item is not returnable.
View return policy
Description
Specifications
ASTM F2413-05 | |
Pull-on | |
16 in. | |
PVC | |
11 | |
Plain |
White | |
Used in commercial fishing, shrimping, and food processing | |
Kick-off | |
Steel | |
White Safety-Loc™ outsole and replaceable cushion insoles, slip-resistant | |
Chemicals |