Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dunlop™ Onguard™ White PVC Boots
$558.00 - $629.74
Specifications
Certifications/Compliance | ASTM F2413-05 |
---|---|
Color | White |
Design | Pull-on |
For Use With (Application) | Used in commercial fishing, shrimping, and food processing |
Height (English) | 16 in. |
Catalog Number | Mfr. No. | Size | Toe Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Toe Style | Price | Quantity & Availability | ||||
19-070731
|
Dunlop™
810110600 |
6 | Plain |
Case of 6 for $558.00
|
|
||||
19-068915
|
Dunlop™
810110800 |
8 | Plain |
Case of 6 for $558.00
|
|
||||
19-068922
|
Dunlop™
810111100 |
11 | Plain |
Case of 6 for $558.00
|
|
||||
19-068917
|
Dunlop™
810110900 |
9 | Plain |
Case of 6 for $558.00
|
|
||||
19-068920
|
Dunlop™
810111000 |
10 | Plain |
Case of 6 for $558.00
|
|
||||
19-068925
|
Dunlop™
810111200 |
12 | Plain |
Case of 6 for $558.00
|
|
||||
19-024270
|
Dunlop™
810120400 |
4 | Steel Toe |
Case of 6 for $603.79
|
|
||||
19-806-582
|
Dunlop™
810120700 |
7 | Steel Toe |
Case of 6 for $603.79
|
|
||||
19-806-581
|
Dunlop™
810120900 |
9 | Steel Toe |
Case of 6 for $603.79
|
|
||||
19-806-583
|
Dunlop™
810120800 |
8 | Steel Toe |
Case of 6 for $603.79
|
|
||||
19-806-586
|
Dunlop™
810121200 |
12 | Steel Toe |
Case of 6 for $603.79
|
|
||||
19-806-585
|
Dunlop™
810121100 |
11 | Steel Toe |
Case of 6 for $603.79
|
|
||||
19-806-587
|
Dunlop™
810121300 |
13 | Steel Toe |
Case of 6 for $603.79
|
|
||||
19-024273
|
Dunlop™
810121000 |
10 | Steel Toe |
Case of 6 for $629.74
|
|
||||
19-070732
|
Dunlop™
810110700 |
7 | Plain | N/A | N/A | N/A | |||
Description
Specifications
ASTM F2413-05 | |
Pull-on | |
16 in. | |
PVC | |
White Safety-Loc™ outsole and replaceable cushion insoles, slip-resistant |
White | |
Used in commercial fishing, shrimping, and food processing | |
Kick-off | |
Steel | |
Chemicals |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title