Onguard White PVC Boots

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 810120800

 View more versions of this product

Catalog No. 19-806-583

  Check Availability
Add to Cart

    Specifications

    Closure Type Pull-up
    Toe Material Steel
    Certifications/Compliance ASTM F2413-05
    For Use With (Application) Used in commercial fishing, shrimping, and food processing
    Gender Male
    Height (English) 16 in.
    Lug Type Kick off
    Resistant Chemical
    Shank Material Steel
    Sole Features White Safety-Loc™ outsole and replaceable cushion insoles, slip resistant
    Toe Style Steel
    Color White
    Material PVC
    Size 8

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit