Onguard White PVC Boots

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 810120400

 View more versions of this product

Catalog No. 19-024270

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-05
    Resistant Chemical
    Closure Type Pull-up
    For Use With (Application) Used in commercial fishing, shrimping, and food processing
    Gender Male
    Height (English) 16 in.
    Lug Type Kick off
    Shank Material Steel
    Toe Material Steel
    Toe Style Steel
    Color White
    Material PVC
    Size 4
    Sole Features White Safety-Loc™ outsole and replaceable cushion insoles, slip resistant

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit