Onguard Women's Plain Toe Standard PVC Boots with Cleated Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 562330800

 View more versions of this product

Catalog No. 19-024259

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Closure Type Pull-up
    Height (English) 14 in.
    Sole Features Tan cleated outsole
    Toe Style Plain
    Color Black
    Size 8
    Certifications/Compliance ASTM F2413-05
    For Use With (Application) Used in general maintenance, construction, and utilities
    Gender Female
    Lug Type Kick off
    Resistant Chemical
    Material PVC

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit