Onguard Women's Steel Toe Standard PVC Boots with Cleated Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 562340500

 View more versions of this product

Catalog No. 19-024265

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-05
    Closure Type Pull-up
    Gender Female
    Height (English) 14 in.
    Lug Type Kick off
    Resistant Chemical
    Sole Features Tan cleated outsole
    Toe Material Steel
    Color Black
    Material PVC
    For Use With (Application) Used in general maintenance, construction, and utilities
    Size 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit