Onguard Women's White PVC Boots

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 510340600

 View more versions of this product

Catalog No. 19-024228

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Height (English) 14 in.
    Lug Type Kick off
    Color White
    For Use With (Application) Used in commercial fishing, shrimping, and food processing
    Certifications/Compliance ASTM F2413-05
    Resistant Mild chemicals
    Toe Material Steel
    Gender Female
    Material PVC
    Seam Type Seamless
    Size 6
    Closure Type Pull-up
    Sole Features Safety-Loc™ outsole and replaceable cushion insoles

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit