Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ONO 1078, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
103177-37-3
C27H23N5O4
481.512
NBQKINXMPLXUET-UHFFFAOYSA-N
pranlukast,azlaire,pranlukast hydrate,onon,pranlukast inn,azlaire tn,n-4-oxo-2-2h-tetrazol-5-yl chromen-8-yl-4-4-phenylbutoxy benzamide,chembl21333,ono-rs-411,4-oxo-8-4-4-phenylbutoxy benzoylamino-2-tetrazol-5-yl-4h-1-benzopyran
4887
N-[4-oxo-2-(2H-tetrazol-5-yl)chromen-8-yl]-4-(4-phenylbutoxy)benzamide
C1=CC=C(C=C1)CCCCOC2=CC=C(C=C2)C(=O)NC3=CC=CC4=C3OC(=CC4=O)C5=NNN=N5
This item is not returnable. View return policy
103177-37-3
C27H23N5O4
481.512
NBQKINXMPLXUET-UHFFFAOYSA-N
pranlukast,azlaire,pranlukast hydrate,onon,pranlukast inn,azlaire tn,n-4-oxo-2-2h-tetrazol-5-yl chromen-8-yl-4-4-phenylbutoxy benzamide,chembl21333,ono-rs-411,4-oxo-8-4-4-phenylbutoxy benzoylamino-2-tetrazol-5-yl-4h-1-benzopyran
4887
N-[4-oxo-2-(2H-tetrazol-5-yl)chromen-8-yl]-4-(4-phenylbutoxy)benzamide
C1=CC=C(C=C1)CCCCOC2=CC=C(C=C2)C(=O)NC3=CC=CC4=C3OC(=CC4=O)C5=NNN=N5

Selective CysLT1 antagonist

Selective cysteinyl leukotriene receptor 1 (CysLT 1 ) antagonist (IC 50 values are ~ 4 - 7 and 3620 nM for CysLT 1 and CysLT 2 respectively). Inhibits contraction of airway smooth muscle, microvascular leakage into airways and eosinophil infiltration. Can decrease symptoms of bronchial asthma.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 481.5
Percent Purity >99%
Chemical Name or Material ONO 1078

Recommended Storage : Store at +4°C