Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ONO 4817, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
223472-31-9
C22H28N2O6
416.474
HDWWQELUBWGQGA-WMZOPIPTSA-N
mmp inhibitor v, d0w8qh, ono-mi1-335, n-hydroxy-2 s-methyl-5-ethoxymethoxy-4 s-n-4-phenoxyphenylcarbonyl amino pentanamide, n-1s,3s-1-ethoxymethoxy methyl-4-hydroxyamino-3-methyl-4-oxobutyl-4-phenoxybenzamide, n-2s,4s-1-ethoxymethoxy-5-hydroxyamino-4-methyl-5-oxopentan-2-yl-4-phenoxy benzamide
9888141
N-[(2S,4S)-1-(ethoxymethoxy)-5-(hydroxyamino)-4-methyl-5-oxopentan-2-yl]-4-phenoxybenzamide
CCOCOCC(CC(C)C(=O)NO)NC(=O)C1=CC=C(C=C1)OC2=CC=CC=C2
This item is not returnable. View return policy
223472-31-9
C22H28N2O6
416.474
HDWWQELUBWGQGA-WMZOPIPTSA-N
mmp inhibitor v, d0w8qh, ono-mi1-335, n-hydroxy-2 s-methyl-5-ethoxymethoxy-4 s-n-4-phenoxyphenylcarbonyl amino pentanamide, n-1s,3s-1-ethoxymethoxy methyl-4-hydroxyamino-3-methyl-4-oxobutyl-4-phenoxybenzamide, n-2s,4s-1-ethoxymethoxy-5-hydroxyamino-4-methyl-5-oxopentan-2-yl-4-phenoxy benzamide
9888141
N-[(2S,4S)-1-(ethoxymethoxy)-5-(hydroxyamino)-4-methyl-5-oxopentan-2-yl]-4-phenoxybenzamide
CCOCOCC(CC(C)C(=O)NO)NC(=O)C1=CC=C(C=C1)OC2=CC=CC=C2

Broad spectrum MMP inhibitor

Broad spectrum MMP inhibitor; Ki values are 0.45, 0.73, 1.1, 1.1, 2.1, 42 and 2500 nM for MMP-12, MMP-2, MMP-8, MMP-13, MMP-9, MMP-3 and MMP-7 respectively and IC50 = 1600 nM for MMP-1). Displays no activity at other proteases up to a concentration of 100 μM. Exhibits antiangiogenic and anti-invasive properties on lung metastasis of murine renal cell carcinoma.
Quantity 10 mg
Formula Weight 416.47
Percent Purity >99%
Chemical Name or Material ONO 4817

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit