Learn More
EVOS™ Vessel Holders and Stage Plates
EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.
Manufacturer:  EVOS™ AMEPVH039
 View more versions of this product
Catalog No. 12-563-606
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EVOS? FL Auto Imaging System
Onstage Vessel Holder, one Lab-Tek II coverglass chamber slide
1 holder
EVOS vessel holder; Dimensions: 128mm x 86mm; Capacity: 1 Nunc LabTech-II Chamber Slide