Learn More
White Knight™ Open Face Reusable Cleanroom Hood
These reusable hoods are suitable for a variety of cleanroom applications including: pharmaceutical, biotechnology, and medical device manufacturing.
$25.25 - $30.75
Specifications
Product Type Reusable
For Use With (Application) To Control The Migration of Particulates in Pharmaceutical Cleanroom Environments
Includes 1 reusable cleanroom hood
Sterility Non-sterile
Material Polyester / Carbon Stripes
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-111-251
View Documents
White Knight™
11HD3262N18001XSWHT
X-Small Each for $25.25
Add to cart
18-111-252
View Documents
White Knight™
11HD3262N18001SWHT
Small Each for $25.25
Add to cart
18-111-253
View Documents
White Knight™
11HD3262N18001MWHT
Medium Each for $25.25
Add to cart
18-111-254
View Documents
White Knight™
11HD3262N18001LWHT
Large Each for $25.25
Add to cart
18-111-255
View Documents
White Knight™
11HD3262N18001XLWHT
X-Large Each for $25.25
Add to cart
18-111-256
View Documents
White Knight™
11HD3262N180012XLWHT
2X-Large Each for $27.75
Add to cart
18-111-257
View Documents
White Knight™
11HD3262N180013XLWHT
3X-Large Each for $30.75
Add to cart
Description
Description
  • Fluid and bacteria resistant Integrity 1800™ high performance carbon striped fabric
  • Open face, split-drape design
  • Laser-cut, pre-serged seams
  • Silicon-free
  • Tunneled elastic plus horizontal and vertical adjustment snaps
Specifications
Specifications
Reusable
1 reusable cleanroom hood
Polyester / Carbon Stripes
Yes
No
To Control The Migration of Particulates in Pharmaceutical Cleanroom Environments
Non-sterile
Integrity 1800
White