Learn More
Humboldt Open Pan Riffle Samplers
Manufacturer:  Humboldt H3962
 View more versions of this product
Catalog No. 04-942A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Sample poured into funnel-shaped trough is divided into two parts by two series of chutes that discharge alternately in opposite directions into separate pans. With four pans, a scoop, and a brush for collecting fine material and cleaning riffles.
Order Info
Available in four chute widths.
Specifications
Specifications
Riffle Sample Splitter
8 in.
20.32cm
8 in.
17.01cm
5.2 in.
0.93cm
4.5 in.
Stainless Steel
Riffle
13.23cm
14
ASTM B215, C136, C702, C778, D421, D424, D457, D806, AASHTO T27, T144, T248
0.37 in.
Divide or halve dry materials such as cement, gravel, powdered ores, coal, coke, sand, soils
4 cold-rolled steel open pans, scoop and a brush
20.32cm
11.43cm
6.7 in.
Small