Thermo Scientific™ Storage Vials & Closures Available on GSA/VA Contract for Federal Government customers only.
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Manufacturer: Thermo Scientific™ B780718
 View more versions of this product
Catalog No. 03-392B
  Check Availability
Add to Cart
    Specifications
    Vial Closure Size 18mm
    Item Description Open top 18-400 Screw Cap
    Material Glass (Vial),Polypropylene (Cap)
    Quantity 100
    Closure Color Black
    Closure Material Polypropylene
    Cap Style Screw Cap (Open-Top)
    Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
    • PTFE-lined solid-top storage caps
    • PTFE on film/foam backing offers broad chemical resistance
    • Not autoclavable
    Order as individual sample storage vials and closures or as unassembled convenience kits.
    Unassembled convenience kits saves time during sample preparation.
    • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
    • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
  • Description & Specifications
Related Products
Product Content Feedback