Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
Open-Top Bell Jars
Made of heavy molded glass.
$205.00
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Price | Quantity & Availability | ||||
S30968
![]() |
Variety Glass
636 1/2 B 6 X 11 |
1 gal. | 3.8 L |
Each for $205.00
|
N/A |
Description
- Ground flange provides seal with pump plate
- Tooled top for a perfect stopper fit
Specifications
Bell Jar | |
Glass |