Learn More
Open-Top Bell Jars Products for Science Education
Made of heavy molded glass.
Manufacturer:  Variety Glass 636 1/2 B 6 X 11
 View more versions of this product
Catalog No. S30968
Description
Description
  • Ground flange provides seal with pump plate
  • Tooled top for a perfect stopper fit
Specifications
Specifications
11 in.
6 in.
3.8 L
1 gal.
Bell Jar
Glass