Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Polypropylene, Platinum-cured Silicone, Open Top Cap with Two 1/4 in. and One 3/8 in. OD Tubes (27 in. Length), 83 mm
For Use With: Compatible on bottles and carboys with 53-mm, 83-mm, or 120-mm closures
Supplier: Justrite™ 12896
Description
- Helps avoid open-air pouring with leak-proof closed system
- Pre-mounted platinum-cured silicone tubing for easy custimization of carboys
Specifications
Two 1/4 in., one 3/8 in. tube, 27 in. L | |
Polypropylene, Polypropylene Cap, Polypropylene Adapter, Platinum-cured Silicone Tubing | |
Compatible on bottles and carboys with 53-mm, 83-mm, or 120-mm closures |
H x Dia.: 45.9 x 5 in. | |
Caps |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title