Learn More
Sklar InstrumentsSupplier Diversity Partner Sklarhone Operating Scissors, Sklar, 5-1/2 in., Sharp/Sharp, Curved, Razor/Smooth, Finger Rings / Dual Black, Stainless Steel. Sklarhone Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 153348
Sklarhone Operating Scissors, Sklar, 5-1/2 in., Sharp/Sharp, Curved, Razor/Smooth, Finger Rings / Dual Black, Stainless Steel. Sklarhone Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Sklarhone scissors feature one knife-like razor sharp edge and one standard scissor edge. Catching skin and pulls are minimized by the use of Sklarhone premium scissors. This product is curved with one knife-like razor sharp edge, one standard scissor edge, sharp/sharp tips, and an overall length of 5-1/2 inches.
Catalog No. 50-143-7374