Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Operating Scissors, Sklar, 5 in., Sharp/Sharp, Straight, Smooth, Finger Rings, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21271
Econo Operating Scissors, Sklar, 5 in., Sharp/Sharp, Straight, Smooth, Finger Rings, Stainless Steel. Econo Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo instruments are, disposable, floor-grade instruments sold in bulk quantities. This product is straight with smooth, sharp/sharp tips and a length of 5 inches. They are sold in boxes of 12.
Catalog No. 50-143-7581