Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ Opti- Tranz™ Liquid Stuart Media Transport Tube with HydraFlock™ Tip Swab
Features three-dimensional microstructure fiber design for sampling
Supplier: Puritan™ LS62H
This item is not returnable.
View return policy
Description
1 ml Liquid Stuart in absorbent polyurethane foam, which acts as a moisture reservoir for bacteriaIntended for collection and transport of aerobic and fastidious bacteria. Used in bacterial culture, rapid tests, and Gram staining. Flocked swab provides superior specimen collection and elution. Double HydraFlock™ swab (3206-H), plastic handle. Shelf Life: 18 Months. Made in USASpecifications
Transport Tube | |
Sterile | |
Screw Cap | |
Polystyrene | |
154 mm |
Semi-solid (Sodium Thioglycollate and Agar) | |
Pink | |
Throat, urogential, and wound swab sampling | |
Double Flock | |
1 mL |
Safety and Handling
ShelfLife : 18 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic (IVD) use only.
Spot an opportunity for improvement?
Provide Content Correction