Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ Opti- Tranz™ Liquid Stuart Media Transport Tube with Miniature Rayon Tip Swab
Ideal for ear, nose, throat, male urethral sampling
Specifications
Product Type | Transport Tube |
---|---|
Media Type | Semi-solid (Sodium Thioglycollate and Agar) |
Sterility | Sterile |
Closure Type | Screw Cap |
For Use With (Application) | Throat, urogential, and wound swab sampling |
Catalog Number | Mfr. No. | Shaft Material | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Shaft Material | Closure Color | Price | Quantity & Availability | ||||
22-027-128
|
Puritan™
LS61AR |
Aluminum | Orange |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
22-027-129
|
Puritan™
LS61SAR |
Soft Aluminum | Green |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
22-027-126
|
Puritan™
LS61PR |
Polystyrene | Red |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
22-027-127
|
Puritan™
LS62PR |
Polystyrene | White |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
Description
- Maintains viability of microorganisms without significant increase in growth
- Non-nutritive general purpose medium
- Sodium thioglycollate as reducing agent, sodium glycerophosphate to buffer
calcium chloride - 1mL sterile liquid stuart gel medium
- Non-toxic rayon tip (Length: 17.45mm)
- Aluminum wire for flexibility and easy shape
Applications
- Aerobic culture
- Ear, nose, throat, and eye sampling, as well as male urethral sampling
- Ideal for direct microscopic examination of clinical samples
Specifications
Transport Tube | |
Sterile | |
Throat, urogential, and wound swab sampling | |
154 mm |
Semi-solid (Sodium Thioglycollate and Agar) | |
Screw Cap | |
Rayon | |
1 mL |
Safety and Handling
ShelfLife : 18 Months
For in vitro diagnostic (IVD) use only.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title