Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ Opti- Tranz™ Plus Liquid Amies Media Transport Tube with HydraFlock™ Tip Swab
Used for isolation of aerobic culture
Specifications
Product Type | Transport Tube |
---|---|
Media Type | Semi-solid (Sodium Thioglycollate and Agar) |
Sterility | Sterile |
Closure Type | Screw Cap |
For Use With (Application) | Throat, urogential, and wound swab sampling |
Catalog Number | Mfr. No. | Tip Type | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Tip Type | Closure Color | Price | Quantity & Availability | ||||
22-027-124
|
Puritan™
LA61H |
Single applicator | Yellow | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
22-027-125
|
Puritan™
LA62H |
Dual applicator | Pink | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Sodium thioglycollate, agar medium
- 1mL sterile amies gel with charcoal
- Aluminum wire for flexibility and easy shape
- Rayon tip (Length: 19.56mm, Dia.: 5.56mm)
- Handle Length: 149.25mm, Dia.: 2.54mm
Applications
- Neutralizes effect of fatty acids in samples that may be toxic to microorganisms
- Ear, nose, throat, eye sampling, as well as male urethral sampling
- Ideal for direct microscopic examination of clinical samples
Specifications
Transport Tube | |
Sterile | |
Throat, urogential, and wound swab sampling | |
Polyester Flock | |
1 mL |
Semi-solid (Sodium Thioglycollate and Agar) | |
Screw Cap | |
Polystyrene | |
154 mm |
Safety and Handling
ShelfLife : 18 Months
For in vitro diagnostic (IVD) use only.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title